missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Feline IgG (H+L) Secondary Antibody, DyLight 594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60651DL594
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Feline samples. It is validated for Flow Cytometry, Immunomicroscopy
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Flow Cytometry : 1:20-1:2000, Immunomicroscopy : 1:20-1:2000 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Flow Cytometry, Immunomicroscopy | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| This Goat anti-Feline IgG (H+L) Secondary Antibody was developed against purified feline IgG (H&L). | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction