Learn More
Goat anti-Human IgA Heavy Chain Secondary Antibody, DyLight 488, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-74908
Product Code. 18516671
  • 373.00€ / 1mg
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgA Heavy Chain
Polyclonal
DyLight 488
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Purified human IgA, (Alpha chain)
1 mg
Adaptive Immunity, Immunology
Rehydrate with 1.1 mL of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily.
Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage.
IgG
Flow Cytometry, Immunomicroscopy
LYOPH
Flow Cytometry 1:20-1:2000, Immunomicroscopy 1:20-1:2000
Goat
Affinity purified
RUO
Secondary