missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgE Heavy Chain Secondary Antibody, PE/Cy7, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74923PECY7
This item is not returnable.
View return policy
Description
IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Specifications
| IgE Heavy Chain | |
| Polyclonal | |
| PBS | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion | |
| PE-Cyanine7 | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Purified human IgE, (Epsilon chain) | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction