missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgG Fc Secondary Antibody, DyLight 755, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74981IR
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
Specifications
| IgG Fc | |
| Polyclonal | |
| Western Blot : 1:100-1:2000, Flow Cytometry : 1:10-1:1000, ELISA : 1:100-1:2000, Immunohistochemistry : 1:10 - 1:500, Immunocytochemistry/Immunofluorescence : 1:10 - 1:500, Immunoassay, Immunomicroscopy : 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA) | |
| O75015, O75015 | |
| Purified Human IgG, Fc fragment | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction