missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human Lambda Light Chain Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72658
This item is not returnable.
View return policy
Description
Lambda Light Chain Polyclonal specifically detects Lambda Light Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Lambda Light Chain | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| IGLC, immunoglobulin lambda constant group | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3536 | |
| Human | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200 - 1:1000 | |
| IGLC1 | |
| Human lambda light chain | |
| 1 mg | |
| Secondary | |
| Specificity was confirmed by ELISA minimal cross reactivity against other human heavy or light chain isotypes. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction