Learn More
Invitrogen™ Goat anti-Mouse IgA Secondary Antibody, AP
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ 626722
This item is not returnable.
View return policy
Description
Goat anti-Mouse IgA Secondary Antibody for ELISA, IHC, Western Blot
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Mouse IgA | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| Goat | |
| Affinity chromatography | |
| RUO | |
| Mouse | |
| Secondary Antibody | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| 0.05M tris with 40% glycerol, 1mM MgCl2, 1% BSA and 0.1% sodium azide; pH 7.5 | |
| Mouse IgA. | |
| 1 mL | |
| Secondary | |
| 4°C | |
| Liquid |