missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgM (H+L) Secondary Antibody, PE/Atto594, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75203PEATT594
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay
Specifications
| IgM (H+L) | |
| Polyclonal | |
| PBS | |
| Purified Mouse IgM, whole molecule | |
| 500 μg | |
| B Cell Development and Differentiation Markers, Immunology | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion | |
| PE/ATTO 594 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction