Learn More
Invitrogen™ Goat anti-Rabbit IgG (Heavy chain) Superclonal™ Secondary Antibody, HRP
Goat Recombinant Superclonal Secondary Antibody
83.75€ - 362.00€
Specifications
| Antigen | Rabbit IgG (Heavy chain) |
|---|---|
| Concentration | 1 mg/mL |
| Content And Storage | 4°C, store in dark, DO NOT FREEZE! |
| Applications | Peptide Array, ELISA, Immunoprecipitation, Western Blot |
| Classification | Recombinant Superclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15306987
|
Invitrogen™
A27036 |
1 mg |
362.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15687868
|
Invitrogen™
A27036SAMPLE |
50 μg |
83.75€
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The sensitivity and specificity of each lot is confirmed using ELISA. Minimal cross-reactivity with mouse, rat, human, bovine, guinea pig and donkey IgG is observed. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and more lot-to-lot consistency.
Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
| Rabbit IgG (Heavy chain) | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Recombinant Superclonal | |
| Liquid | |
| IgG | |
| Rabbit | |
| Recombinant full-length protein. | |
| Secondary Antibody |
| 1 mg/mL | |
| Peptide Array, ELISA, Immunoprecipitation, Western Blot | |
| HRP | |
| Goat | |
| RUO | |
| PBS with HRP Stabilizer and proprietary preservative | |
| Secondary | |
| Gravity column chromatography |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.