Learn More
Epredia™ Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Protect and preserve specimens on slides with Epredia™ Gold Seal™ Cover Glasses, made of noncorrosive borosilicate glass.
16.95€ - 21.20€
Product Code | Brand | Shape | Thickness (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Shape | Thickness (Metric) | Dimensions (L x W) | Price | Quantity & Availability | |||
15809814
|
Epredia™
22X22-1-002 |
Square | #1 (0.13 to 0.16 mm) | 22 x 22 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
15819814
|
Epredia™
22X22-1.5-002 |
Square | #1.5 (0.16 to 0.19 mm) | 22 x 22 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
15829814
|
Epredia™
22X22-2-002 |
Square | #2 (0.19 to 0.23 mm) | 22 x 22 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||
Description
Each case of 10 ounces converts into a convenient 100-slide storage file with a protective clear cover. Applications include hematology, histology, cytology, microbiology, and urology. Available in a variety of thicknesses and sizes.
- Virtually colorless appearance
- Very low levels of inclusions, striae, bubbles, streaks
- High special transmission
- Finely adaptive refractive index suited for microscopes
- Low auto-fluorescence and high chemical resistance
- Made of noncorrosive borosilicate glass
- Sealed in a moisture-resistant plastic case with desiccant packs