Learn More
Epredia™ Gold Seal™ Cover Glasses, rectange, 40 x 22mm, Thickness 1
Protect and preserve specimens on slides with Epredia™ Gold Seal™ Cover Glasses, made of noncorrosive borosilicate glass.
Brand: Epredia™ 22X22-1.5-002
Additional Details : Weight : 0.28000kg
Description
Each case of 10 ounces converts into a convenient 100-slide storage file with a protective clear cover. Applications include hematology, histology, cytology, microbiology, and urology. Available in a variety of thicknesses and sizes.
- Virtually colorless appearance
- Very low levels of inclusions, striae, bubbles, streaks
- High special transmission
- Finely adaptive refractive index suited for microscopes
- Low auto-fluorescence and high chemical resistance
- Made of noncorrosive borosilicate glass
- Sealed in a moisture-resistant plastic case with desiccant packs
Specifications
Square | |
22 mm | |
#1.5 (0.16 to 0.19 mm) | |
Approx. 140/bx., 1400/cs. |
Borosilicate Glass | |
22 mm | |
22 x 22 mm |