missing translation for 'onlineSavingsMsg'
Learn More

Gold wire, 0.762mm (0.030in) dia, Premion™, 99.999% (metals basis)

Product Code. 11388827
Click to view available options
Quantity:
250 cm
Unit Size:
250cm
This item is not returnable. View return policy

Product Code. 11388827

Brand: Thermo Scientific Alfa Aesar 038379.CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Form: ≈8.8g/m | CAS: 7440-57-5

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-57-5
Quantity 250 cm
Percent Purity 99.999%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.