Learn More
Corning™ Octagonal Polyethylene Terephtalate Bottles
Catalog Number 16257280
Click to view available options
Capacity:
16.9 oz., 500 mL
33.8 oz., 1000 mL
4.2 oz., 125 mL
8.45 oz., 250 mL
Closure Color:
Natural
Packaging:
Bulk
Retractable Film Packed
Tray Filmed Packed
Designed with a leak-resistant closure system
  • Break-resistant polyethylene terephthalate (PET) with transparency equivalent to glass
  • Molded graduations
  • Tamper-evident High-density Polyethylene (HDPE) cap
  • Film-packed with or without tray in folded outer bag
TRUSTED_SUSTAINABILITY
Capacity 4.2 oz., 125 mL
Closure Material HDPE
Closure Type Screw Cap
Diameter (Metric) 53 mm
Height (English) 4.1 in.
Height (Metric) 105 mm
Capacity (Metric) 125 mL
Capacity (English) 4.2 oz.
Color Clear
Diameter (English) 2 in.
Material PET
Length (Metric) 53 mm
Packaging Retractable Film Packed
Sterility Sterile
Width (Metric) 53 mm
Closure Color Natural
Graduations 25 mL
Certifications/Compliance ISO 9001:2008/ISO 11137 : STERILE SAL 10-3
Autoclavable Not Autoclavable