Learn More
Gosselin™ Self-Standing Conical Tube, PP
Suitable for centrifugation and conveniently graduated conical tube. Gosselin™ Self-Standing Conical Tube, PP is conically shaped, but has a flat bottom to facilitate self standing.
Brand:  Gosselin™ TU50-012
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15281852
  • 105.00€ / Pack of 500
Estimated Shipment
08-07-2020
Add to basket
Description
Description
  • Durable polypropylene construction
  • Conical with a flat bottom
  • Printed graduations
  • Green cap
Specifications
Specifications
29.2mm
Screw cap
Polypropylene
500/Cs
Non-sterile
Green
Centrifuge
50/Pk