Learn More
Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth
Product Code. 10339204
Click to view available options
Diameter (Metric) Outside:
32 mm
47 mm
Unit Size:
Pack of 100
Product Code. 10339204
Brand: Poeppelmann™ 60004130000
Only null left
Add to basket
Add to basket
Excellent for closing off bore holes and internal threads, or to protect studs and external threads. Poeppelmann™ KAPSTO™ GPN 600 Tapered Closures, 14mm Depth are manufactured from natural-colored PE-LD plastic.
  • 14mm insertion depth
  • Universal use as plug or cap
  • Tapered design ideal for accomodating tolerances
  • Chemically resistant to alcohol and weak acids/alkalis
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 14 mm
Quantity 100 Pack
Size (Metric) O.D. x H: 47 x 14 mm, 14 mm Insertion Depth
Diameter (Metric) Outside 47 mm
Material LDPE
Type Tapered Closure