missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GR 55562 dihydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
A selective competitive 5-HT1B (5-HT1Dβ) silent antagonist with pKB values of 7.3 and 6.3 for human cloned 5-HT1B and 5-HT1D receptors respectively and only weak binding at a number of other 5-HT subtypes.
Specifications
Quantity | 50 mg |
Formula Weight | 448.39 |
Percent Purity | >98% |
Chemical Name or Material | GR 55562 dihydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'