missing translation for 'onlineSavingsMsg'
Learn More

GR 89696 fumarate, Tocris Bioscience™

Catalog Number 18044722
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
126766-32-3
C23H29Cl2N3O7
530.399
ABTNETSDXZBJTE-WLHGVMLRSA-N
gr 89696 fumarate, gr-89696 fumarate, gr 89696 fumarate salt, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate salt, 1-3,4-dichlorophenyl acetyl-4-acetyl-2-1-pyrrolidinylmethyl piperazine, methyl 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylate fumarate, gr 89696 fumarate salt, solid, 4-3,4-dichlorophenyl acetyl-3
6442840
(E)-but-2-enedioic acid;methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-(pyrrolidin-1-ylmethyl)piperazine-1-carboxylate
COC(=O)N1CCN(C(C1)CN2CCCC2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.C(=CC(=O)O)C(=O)O
This item is not returnable. View return policy
126766-32-3
C23H29Cl2N3O7
530.399
ABTNETSDXZBJTE-WLHGVMLRSA-N
gr 89696 fumarate, gr-89696 fumarate, gr 89696 fumarate salt, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate, 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylic acid methyl ester fumarate salt, 1-3,4-dichlorophenyl acetyl-4-acetyl-2-1-pyrrolidinylmethyl piperazine, methyl 4-3,4-dichlorophenyl acetyl-3-1-pyrrolidinylmethyl-1-piperazinecarboxylate fumarate, gr 89696 fumarate salt, solid, 4-3,4-dichlorophenyl acetyl-3
6442840
(E)-but-2-enedioic acid;methyl 4-[2-(3,4-dichlorophenyl)acetyl]-3-(pyrrolidin-1-ylmethyl)piperazine-1-carboxylate
COC(=O)N1CCN(C(C1)CN2CCCC2)C(=O)CC3=CC(=C(C=C3)Cl)Cl.C(=CC(=O)O)C(=O)O

Subtype-selective κ agonist

Highly potent and selective κ-opioid agonist (IC50 = 0.04 nM) that may be selective for the putative κ2 receptor. Anti-nociceptive and neuroprotective in vivo.
Quantity 50 mg
Formula Weight 530.4
Percent Purity >99%
Chemical Name or Material GR 89696 fumarate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'