Learn More
Sartorius™ Grade 10 Smooth Surface Qualitative Filter Papers
Used for analytical purposes and routine analyses.
Brand:  Sartorius™ FT-2-352-580580
Additional Details : Weight : 0.01000kg
Product Code. 10684522
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Used for analytical purposes and routine analyses.
Specifications
Specifications
Sheet
7.5μm/sec.
10
0.33 mm
580 L x 580 Wmm
580 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100 Pack
Qualitative Filter Paper
580 mm