Learn More
Fisherbrand™ Grade 1100 Cellulose Technical Filter Paper
For filtration of coarse particles and gelatinous precipitates.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11729955
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Creped thin paper
  • Wet strengthened
  • For filtering viscous liquids as arabic gum, gelatine, emulsions like agar-agar, alginate
Specifications
Specifications
Disc
50 to 60 μm
Viscous liquids as Arabic gum, Gelatin, emulsions like Agar-agar, Alginate, protective Paper for filter presses
100 Pack
185 mm
Cellulose
50 to 60 μm
1100
0.2 mm
185 mm