Learn More
Fisherbrand™ Grade 1106 Cellulose Technical Filter Paper
Used for viscous liquids and coarse particles.
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11780026
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Thick smooth paper with large pore structure
Specifications
Specifications
Disc
50 to 80 μm
For syrups, animal fats, vegetable fats, margarines, Gelatin, dense oils, emulsions, essential oils
100 Pack
185 mm
Cellulose
50 to 80 μm
1106
0.47 mm
185 mm