Learn More
Fisherbrand™ Grade 1107 Cellulose Technical Filter Paper: Discs
Used for filtration of semi fine particles
Brand:  Fisherbrand™
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11750056
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
  • Thick smooth paper with medium pore structure
Specifications
Specifications
Folded Disc
20 to 30 μm
For Serum, Culture Medium, dyes, tinctures, vegetal extracts, turbid liquids, juices, Eaux de Cologne, perfumes, essences, hair tonics
100 Pack
150 mm
Cellulose
20 to 30 μm
1107
0.38 mm
150 mm