missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Grade 50 S, Smooth Papers for Qualitative and Technical Analyses, Discs

Sartorius Stedim Biotech™ Grade 50 S, Smooth Papers for Qualitative and Technical Analyses, Discs are analytical and technical papers with a medium fast filtering rate, and can be used as termination test paper for the TP method.

Brand:  Sartorius Stedim Biotech™ FT-3-353-095

Additional Details : Weight : 1.00000kg

Product Code. 15972930

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »


Description

Description

  • Made of refined pulp and cotton linters with > 95% alpha-cellulose
  • Medium fast filtering
  • Non-sterile, bulk-packed
Specifications

Specifications

120 g/m2
Cotton Linters, Refined Pulp
30 sec.
50/Pk.
Qualitative Filter Paper
Disc
6 to 10 μm
50 S
0.22 μm
95 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions