Learn More
Fisherbrand™ Grade 912 Universal Test Paper
Product Code. 10754213 Shop All Fisherbrand Products
Click to view available options
Type:
Blue Litmus Paper
KI Starched Paper
Neutral Litmus Paper
Red Litmus Paper
Color Indicator:
Blue
Purple
Red
White
Dimensions (L x W):
5 m x 10 mm
5 m x 7 mm
60 x 10 mm
Format:
Roll
Tube
Unit Size:
Each
Pack of 200
Pack of 4
Product Code. 10754213
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Used for general analysis.
  • General control of acid or alkaline reactions
Color:
  • Litmus Blue: Blue to red
  • Litmus neutral: Purple to blue/red
  • Litmus red: Red to blue
Applications:
  • Rapid determination of pH value in industrial applications
  • General analysis in school
  • Used in formation
Specifications
Color Indicator Red
Format Roll
For Use With (Application) Rapid determination of pH value in industrial applications, general analysis in school, formation
Quantity 1 Roll
Type Red Litmus Paper
Dimensions (L x W) 5 m x 10 mm
Length (Metric) 5 m
Width (Metric) 10 mm