Learn More
Kartell™ Plastilab™ Series 300 Graduated Narrow Neck Bottles
Feature double-sealing with insert cap to ensure tightness.
Brand:  Kartell™ 0032300
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12681006
  • 493.00€ / Pack of 300
Estimated Shipment
19-03-2021
Add to basket
Description
Description
  • Made from polyethylene
  • Sloping shoulders reduce tendency for residue deposits
  • No leaking with easy-grip cap
  • Tagging points on shoulder and cap ensure tamper-proof closure
  • Suitable for storing food stuff
Specifications
Specifications
Regulation 1935/2004, EC Regulation 10/2011
Narrow
LDPE
Cylinder Bottle
Insert Cap
250 mL
23mm
25mL