Learn More
Vitlab™ Graduated Stacking Pitchers
Catalog Number 15362964
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Color:
Green
Red
Yellow
Measure and carry-out chemical reactions. Vitlab™ Graduated Stacking Pitchers are ideal for performing chemical reactions in the laboratory and measuring chemicals. It has a stable handle and easily readable, printed scale on both sides.
  • Available in four different colors
  • With stable handle and easily readable, printed scale on both sides
  • Clean at no higher than 60 °C is recommended
Color Yellow
Diameter (Metric) Inner 100 mm
Capacity (Metric) 500 mL
Material Polypropylene
Graduation Range 10 mL
Height (Metric) 140 mm
Quantity 12 Pk.
Autoclavable Autoclavable
Graduated Yes