Learn More
Grant Instruments™ Grant Bio™ Replacement Rotors
Product Code. p-8013363
Click to view available options
For Use With (Equipment):
12 x 0.5 mL, 12 x 0.2 mL Microtubes
12 x 1.5 mL Microtubes
Unit Size:
Each
Product Code. 15211236
Brand: Grant Instruments™ R0502
Includes microtubes. For use with PVC Series Benchtop Centrifuges
For use with PVC Series Benchtop Centrifuges
  • Includes microtubes
  • R-05-02: Replacement rotor for 12 x 0.5mL and 12 x 0.2mL microtubes
  • R-15: Replacement rotor for 12 x 1.5mL microtubes
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor
For Use With (Equipment) 12 x 0.5 mL, 12 x 0.2 mL Microtubes