Learn More
Trajan Scientific and Medical™ Replacement Mini Union Ferrules
Product Code. 12736348
Click to view available options
For Use With (Equipment):
0.1 to 0.32 mm I. D Columns
0.45 to 0.53 mm ID Columns
Unit Size:
Pack of 10
Product Code. 12736348
Brand: Trajan Scientific and Medical™ 072629
Only null left
Add to basket
Add to basket
Leak-free system connections for improved chromatography for non-MS systems
  • Easy to use
  • Forms a reliable seal
  • Reusable
  • Soft material, forms a soft grip to the capillary column
  • Low emissions
Specifications
Type Ferrule
For Use With (Equipment) 0.45 to 0.53 mm ID Columns
Material Graphite
Quantity 10 Pack