Learn More
Trajan Scientific and Medical™ Replacement Mini Union Ferrules
Leak-free system connections for improved chromatography for non-MS systems
Brand: Trajan Scientific and Medical™ 072630
Additional Details : Weight : 0.01000kg
Description
- Easy to use
- Forms a reliable seal
- Reusable
- Soft material, forms a soft grip to the capillary column
- Low emissions
Specifications
0.1 to 0.32 mm I. D Columns | |
Graphite | |
Ferrule | |
10 Pack |