Learn More
Graphite plate, highly oriented pyrolytic graphite (HOPG), 10x10x1mm (0.394x0.394x0.039in), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Pc.
Packaging:
Shrink-wrapped
Unit Size:
Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Chemical Name or Material | Graphite plate |
| Packaging | Shrink-wrapped |
| MDL Number | MFCD00144065 |
| Quantity | 1 Pc. |