Learn More
Olympus™ Graticule, Crosshair
Product Code. 12582347
Click to view available options
Type:
Crosshair Eyepiece Micrometer
Unit Size:
Each
Product Code. 12582347
Brand: Olympus™ K10017196
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Make accurate measurements of specimens with this eyepiece micrometer. Olympus™ Graticule, Crosshair is a crosshair-type micrometer glass-engraved with a 10mm line with 0.1mm divisions on the X-axis.
  • Glass-engraved.
  • Crosshair-type.
  • X-axis measuring
Specifications
Type Crosshair Eyepiece Micrometer
Description 10mm line, 0.1mm divisions on X-axis
For Use With (Equipment) WHN10X, WHN10X-H, WH10X, WH10X-H, WH15X, WHS10X-H, WHS15X-H, CWH10X, CWH10X-H