Learn More
Ansell™ Comfort G26G Nitrile Immersion Gloves
Product Code. 15314552 Shop All Ansell Products
Click to view available options
Size:
6.5
7.5
8.5
10.5
Unit Size:
12 pairs
Product Code. 15314552
Brand: Ansell™ A340065
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Excellent resistance to many solvents and oil based chemicals. Flexible and comfortable for an excellent dexterity. Superior raised grip pattern for safety handling.
  • Very good abrasion resistance.
  • Allows contact up to 100°C.
  • Superior raised grip pattern for safety handlings.
  • Open cuff with textile edge binding allows ventilation to keep hand cool.
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nitrile
Cuff Material Nitrile
Product Type Immersion Gloves
Size 6.5
Glove Cuff Color Green
Glove Cuff Style Gauntlet
Liner Material Cotton
Model COMFORT G26G
Quantity 12 Pairs
Resistant To Excellent resistance to many solvents and oil based chemicals