Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
Product Code. 15402560
Click to view available options
Capacity (Metric):
2.5 mL
3.5 mL
5 mL
8 mL
13 x 100 mm
Closure Color:
Gold
Red with White Ring
Red with Yellow Ring
Unit Size:
Pack of 100
Pack of 1200
Pack of 50
Product Code. 15402560
Brand: Greiner Bio-One 456073
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
For routine clinical chemistry, immunohematology, serology or immunology testing
  • Coated with micronized silica particles which activate clotting when tubes are gently inverted
  • Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Pediatric tubes come with white rings for quick identification
  • Safety Twist Caps can be removed and replaced several times
  • Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
TRUSTED_SUSTAINABILITY
Specifications
Additive Microscopic Silica Particles with Separation Gel
Material Plastic
Closure Type Non-ridged Pull Cap
Capacity (Metric) 13 x 100 mm
Closure Color Red with Yellow Ring