Learn More
Greiner Bio-One VACUETTE™ Serum Tubes
Product Code. p-3240862
Click to view available options
Capacity (Metric):
5 mL
Unit Size:
Pack of 50
Product Code. 15492550
Brand: Greiner Bio-One 456071
With safety twist cap. The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
  • Inner wall lining: microscopic silica particles
  • Contain separation gel in base of tube
  • Cap color: red
  • Ring color: yellow
  • With Z Serum Sep Clot Activator
TRUSTED_SUSTAINABILITY
Specifications
Additive CAT Serum Sep Clot Activator
Sterility Sterile
Capacity (Metric) 5 mL
Closure Color Red
Length (Metric) 100 mm
Diameter (Metric) Outer 13 mm