Description
- Chemically inert
- Angle-cut end help shear the sample matrix and ensure good homogenisation and maximise extraction rates
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Dimensions (L x W) | 3/8 x 7/8 in. |
| For Use With (Application) | Two cylinders are used per sample vial |
| Packaging Quantity | 100/bag |