Learn More
Microspec™ Grip Seal Bags
Product Code. 15792175
Click to view available options
Dimensions (L x W):
2 x 9 in.
3 x 7.5 in.
6 x 9 in.
7.5 x 7.5 in.
Unit Size:
Pack of 1000
Product Code. 15792175
Brand: Microspec™ W4
Only null left
Add to basket
Add to basket
Provides storage for large number of samples
  • Cost effective sampling
  • Sample storage of large numbers of samples
Specifications
Content And Storage Room Temperature
Autoclavable Not Autoclavable
Closure Type Grip Seal
Color Transparent
Disposable Single-use
For Use With (Application) Sampling
Material Polyethylene
Sterility Non-sterile
Thickness (Metric) 50 μm
Film Description Polyethylene
No. per Case 1000
No. per Pack 1000
Outer Packaging Box
Quantity 1000
Dimensions (L x W) 7.5 x 7.5 in.
Product Type Sample Bag