missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Chicken Thyroxine ELISA Kit (Colorimetric)

Product Code. 18222485 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18222485

Brand: Novus Biologicals™ NBP2601511KIT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High sensitivity and specificity for detection of chicken T4.

The Chicken Thyroxine ELISA Kit (Colorimetric) measures chicken Thyroxine in serum, plasma.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 20 to 320ng/mL
Assay Sensitivity 20ng/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum
Specificity For detection of chicken T4
Target Species Chicken
For Use With (Application) ELISA
Kit Contents Instruction manual, Standard, Stop Solution, Wash Buffer (20 x concentrate), Substrate B, Substrate A, HRP-avidin, Conjugate, Assay plate
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Signal Transduction
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Synonym T4
Target Thyroxine
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.