missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GSG2 (Haspin) Recombinant Human Protein
GSG2 phosphorylates histone H3 and is viewed as a potential therapeutic target for oncology.
602.00€ - 7440.00€
Specifications
Accession Number | NP_114171.1 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. |
Molecular Weight (g/mol) | 64.8kDa |
Storage Requirements | Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Gene Alias | GSG2 (Haspin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10778185
|
Invitrogen™
PV5708 |
10μg |
602.00€
Each |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15408713
|
Invitrogen™
PR9069A |
100μg |
1722.00€
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15448793
|
Invitrogen™
PV5709 |
1mg |
7440.00€
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
GSG2 (haspin) is a conserved kinase required for mitosis.
Specifications
NP_114171.1 | |
64.8kDa | |
GSG2 (Haspin) | |
Recombinant | |
Baculovirus |
50mM Tris pH 7.5, 150mM NaCl, 0.02% Trition X-100, 0.5mM EDTA, 2mM DTT, 50% Glycerol. | |
Kinase proteins are shipped on dry ice. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Human | |
GST Tag |