missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GSK 1016790A

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GSK 1016790A is a potent TRPV4 agonist; elicits Ca2+ influx in human and mouse TRPV4-expressing HEK cells (EC50 values of 2.1 and 18 nM, respectively). Enhances insulin mRNA expression. Increases ERK1/2 phosphorylation and NO production. Exhibits no activity at TRPM8 and TRPA1 (20 μM) channels. Induces bladder overactivity in TRPV4+/+ mice.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C28H32Cl2N4O6S2 |
| Formula Weight | 655.61 |
| Product Type | Agonist |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction