missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 1016790A
GREENER_CHOICE

Product Code. p-7166780
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18750538

Brand: Tocris Bioscience™ 6433/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TRPV4 agonist

GSK 1016790A is a potent TRPV4 agonist; elicits Ca2+ influx in human and mouse TRPV4-expressing HEK cells (EC50 values of 2.1 and 18 nM, respectively). Enhances insulin mRNA expression. Increases ERK1/2 phosphorylation and NO production. Exhibits no activity at TRPM8 and TRPA1 (20 μM) channels. Induces bladder overactivity in TRPV4+/+ mice.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C28H32Cl2N4O6S2
Formula Weight 655.61
Product Type Agonist
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.