missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK J5 HCl
GREENER_CHOICE

Product Code. 18766526
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18766526

Brand: Tocris Bioscience™ 6935/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inactive isomer of GSK J4; cell permeable

GSK J5 HCl is an inactive isomer of GSK J4 (Cat. No. 4594); also cell permeable ester derivative of the inactive control, GSK J2 (Cat. No. 4688).

Active Analog also available.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Molecular Formula C24H27N5O2.HCl
Formula Weight 453.97
Product Type GSK J5
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.