Learn More
Invitrogen™ GSK3 beta (Total/Phospho) Multispecies InstantOne™ ELISA Kit
Description
Includes
- User Manual
- InstantOne™ Assay Plate - 96 well (sealed)
- Cell Lysis Buffer (5X)
- Enhancer Solution
- Wash Buffer (10X)
- GSK3b (Total) Capture Antibody Reagent
- phospho-GSK3b (Ser9) Capture Antibody Reagent
- GSK3b (Total) Detection Antibody Reagent
- phospho-GSK3b (Ser9) Detection Antibody Reagent
- Positive Control Cell Lysate (lyophilized)
- Detection Reagent (TMB Substrate)
- Stop Solution
- Plate Seal
The InstantOne™ ELISA is specifically engineered for accurate measurement of total and phosphorylated human and mouse GSK3b (Ser9) in cell lysates. Detection of total and phosphorylated rat GSK3b (Ser9) is also expected. The InstantOne™ ELISA kit allows for fast analysis of samples in approximately one hour. All reagents used in a traditional sandwich ELISA are added in solution to a plate followed by a wash step and detection with the TMB colorimetric substrate.
Specifications
Specifications
| Accession Number | P49841,Q9WV60 |
| Conjugate | Biotin |
| Product Type | ELISA Kit |
| Sample Type | Cell Lysates |
| Target Species | Human, Mouse |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 2932, 56637 |
| Gene Symbol | Gsk3b |
| Kit Contents | Capture Antibody, Detection Antibody Reagent, InstantOne™ Assay Plate, Cell Lysis Buffer, Enhancer Solution, Wash Buffer (10X), Substrate Reagent (TMB), Stop Solution, Positive Control Cell Lysate (lyophilized), Plate seal |
| Show More |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.