missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ GT 949

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
GT 949 is a potent and selective EAAT2 positive allosteric modulator (EC50 = 0.26 nM). Enhances the glutamate translocation rate, with no effect on substrate interaction. Enhances glutamate uptake by cultured astrocytes by approximately 58%. Exhibits no significant effect on DAT, SERT and NET or NMDA receptors. Neuroprotective in an in vitro model of excitotoxicity.
Specifications
Specifications
| Chemical Name or Material | 3-((4-Cyclohexylpiperazin-1-yl)(1-phenethyl-1H-tetrazol-5-yl)methyl)-6-methoxyquinolin-2(1H)-one |
| CAS | 460330-27-2 |
| Quantity | 10 mg |
| Molecular Formula | C30H37N7O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction