Learn More
Hamilton™ 800 Series Microliter™ Replacement Syringe Barrels
Click to view available options
For Use With (Equipment):
801 N Syringe
801 RN Syringe
802 RN Syringe
810 RN Syringe
8805 RN Syringe
8810 RN Syringe
8825 RN Syringe
Includes:
With needle (22s/51/2)
With needle (26s/51/2)
Without needle
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
Unit Size:
Each
Specifications
Specifications
| For Use With (Application) | Liquids |
| Includes | With needle (22s/51/2) |
| Product Type | Barrels/Plunger Assembly with Needle |
| Volume (Metric) | 100 μL |
| For Use With (Equipment) | 810 RN Syringe |