Learn More
Hamilton™ 800 Series Microliter™ Replacement Syringe Barrels
Product Code. 10759311
Click to view available options
For Use With (Equipment):
801 N Syringe
801 RN Syringe
802 RN Syringe
810 RN Syringe
8805 RN Syringe
8810 RN Syringe
8825 RN Syringe
Includes:
With needle (22s/51/2)
With needle (26s/51/2)
Without needle
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
250 μL
Unit Size:
Each
Product Code. 10759311
Brand: Hamilton™ 32166
Barrels available with or without needle. Choose removable needle (RN) or cemented needle (N) termination. (See exact specifications below.)
Specifications
For Use With (Application) Liquids
Includes With needle (22s/51/2)
Product Type Barrels/Plunger Assembly with Needle
Volume (Metric) 25 μL
For Use With (Equipment) 802 RN Syringe