Learn More
Hamilton™ Chaney Adapter Assemb 700 800 7000/1700 1800 Series Microliter/Gastight™ Syring
Product Code. 10015600
Click to view available options
For Use With (Equipment):
Hamilton Syringe Models 702-750, 7000 and 1700 Series
Hamilton Syringe Models 75 and 701 Series
Hamilton Syringe Models 800 and 1800 Series
Unit Size:
Each
Product Code. 10015600
Brand: Hamilton™ 14725
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Designed to work with 700, 800, 7000 series Microliter syringes and 1700 and 1800 series Gastight syringes
Adapters are designed to give consistent and reproducible injections and also prevent the plungers from bending.
Specifications
Product Type Reproducibility Adapter
Quantity Ea.
For Use With (Equipment) Hamilton Syringe Models 702-750, 7000 and 1700 Series