Learn More
ATAGO™ Handheld Refractometer Master-PM
Master-PM Refractometer can be used for measuring a variety of sauces, soy sauce, seasoning in liquid form as well as for low-sugar jams and spreads.
Brand:  ATAGO™ 2393
Additional Details : Weight : 0.10000kg
Product Code. 11436148
  • 238.00€ / Each
Estimated Shipment: 04-07-2024
to see stock.
Add to basket
Description
Description
  • Can be widely used for beverages, fruit juices, soups, beef bowl sauce, water-soluble cutting oil, etc.
  • Master-PM is made of plastic.
  • For use with salty and acidic samples.
  • This product is the cheapest type with no temperature compensation function.
Specifications
Specifications
Portable Refractometer
MASTER-PM
Brix
32 x 34 x 203 mm
0.0% to 33.0%
0.2% (Brix)
105 g