Learn More
RETSCH Hardened Stainless Steel and Tungsten Carbide O-ring
Shop All Retsch Products
Click to view available options
For Use With (Equipment):
Planetary Ball Mill PM 100 CM 125mL Grinding Jars
Planetary Ball Mill PM 100 CM 250mL Grinding Jars
Planetary Ball Mill PM 100 CM 50mL Grinding Jars
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Planetary Ball Mill PM 100 CM 250mL Grinding Jars |
| Quantity | 1Each |
| Product Type | O-Ring |
| Material | Hardened Stainless Steel and Tungsten Carbide |