Learn More
RETSCH Hardened Steel Mortar
Mix and homogenize powders, suspension and pastes even with high viscosity. Retsch™ Hardened Steel Mortars are suitable for the proper and reproducible sample preparation to analytical fineness.
Brand: RETSCH
Additional Details : Weight : 3.50000kg
Description
- For use with RM 200 Mortar Grinder
Specifications
Mortar | |
Hardened Steel | |
Mortar Grinder RM 200 | |
Mortar |