missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Hardness Reagent Sets, For Colorimetric Water Testing
Click to view available options
Range:
0 to 250 mg/L as CaCO3
0 to 250 mg/L, 200 to 500 mg/L, 400 to 750 mg/L as CaCO3
0 to 400 mg/L Calcium, 0 to 150 mg/L Magnesium
0.00 to 2.70 mg/L
200 to 500 mg/L as CaCO3
No. of Tests:
100
100 (50 Ca/50 Mg)
300
For Use With (Application):
Calcium Hardness
Calcium and Magnesium Hardness High
Total Hardness Low
Total Hardness Low, Medium, High Range
Total Hardness Medium Range
Unit Size:
Pack of 100
Pack of 300
Description
These high quality reagents are designed to follow accepted test methods for colorimetric determination of various parameters of water hardness. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets are available for determination of:
- Calcium Hardness (using liquid or powdered reagents)
- Calcium and Magnesium Hardness (high range)
- Magnesium and Total Hardness
- Total Hardness (low, medium and/or high range)
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Specifications
| Analytes | Calcium: Oxalate method, Magnesium: Calmagite method |
| For Use With (Application) | Calcium and Magnesium Hardness High |
| For Use With (Equipment) | Compatible Hanna benchtop and portable photometers |
| Includes | Dropper bottles |
| No. of Tests | 100 (50 Ca/50 Mg) |
| Quantity | 1 Set |
| Range | 0 to 400 mg/L Calcium, 0 to 150 mg/L Magnesium |
| Test Method | Cholorimetric determination |
| Type | Water Testing Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction