Learn More
Thermo Scientific™ Harvest RC, BC340
Code produit 31767526 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Type:
Chromatography Filter
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type Conditionnement
31767526 Chromatography Filter 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 31767526 Fournisseur Thermo Scientific™ Code fournisseur 7100236865
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
missing translation for 'validMainframeMsg'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
The Harvest RC Chromatographic Clarifier is a single stage single-use chromatographic clarification solution designed for harvesting and clarification of modern cell cultures. Clarification with this solution offers fewer processing steps, higher yield and lower production time and costs.
  • Simplification of high cell density cell culture fluid clarification unit operations.
  • Remove cells, cell debris and DNA
  • Optimized for PCV high density CHO cell culture (5-8% PCV)
  • Replacement of primary, secondary and guard membrane clarification stages.
  • Typical product recovery of 95+% (capsules)
  • Synthetic chromatographic harvest media with chemically defined extractables.
  • Predictable scaling from discovery to manufacturing.
  • Lower total cost of manufacturing compared to centrifugation and depth filtration.
  • Lower consumption of buffer and water compared to depth filtration.
  • Capsules fit into laboratory to manufacturing scale workflows.
Spécification
Jetable Single-use
Type de filtre Filter Capsule
Matériau (membrane) PES
Stérilité Non-sterile
Quantité 1 Ea.
Type Chromatography Filter