Learn More
Brady™ Hazardous Chemical Labels for B30 Series
Product Code. 15921011
Click to view available options
Display:
L3B Layout
L4A Layout
Unit Size:
Pack of 200
Product Code. 15921011
Brand: Brady™ 121648
Only null left
Add to basket
Add to basket
Pre-printed, GHS/CLP chemical labels for BBP3X, S3XXX, and i3300 printers.
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
Specifications
Autoclavable Not Autoclavable
Adhesive Type Permanent Acrylic
Color Red on White
Display L3B Layout
Finish Gloss
Material Vinyl (B-7569)
Print Technology Thermal Transfer
Sterility Non-sterile
For Use With (Application) Facility Identification, Hazard Communication Identification, Electrical Labeling
For Use With (Equipment) BBP™30, BBP™31, BBP™33, BBP™35 Multicolor, BBP™37 Multicolor, BradyPrinter i3300, BradyPrinter S3000, S3100, and i5300 Industrial Label Printers
Labels per Row 1
Quantity 200 Labels
Shape Rectangle
Temperature Range (Metric) -20°C to +75°C
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 162 mm
Width (Metric) 155 mm
Height (Metric) 105 mm
Alternate Part Number B30-262-7569-CLP3B